1,4-Bis(methoxydimethylsilyl)benzene 97%

Supplier: Thermo Scientific Chemicals
Warning

H28165-03 H28165-06
AAH28165-03EA 41.1 USD
AAH28165-03 AAH28165-06
1,4-Bis(methoxydimethylsilyl)benzene 97%
1,4-Bis(methoxydimethylsilyl)benzene

Purity: 97% CAS No.: 131182-68-8 Molecular Formula: C12H22O2Si2 Formula Weight: 254.47 MDL No.: MFCD09266162

Formula: C₁₂H₂₂O₂Si₂
MW: 254.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09266162
CAS Number: 131182-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR