1-Benzyloxycarbonylpyrrolidine-3-carboxaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Benzyl 3-formylpyrrolidine-1-carboxylate
H28117-03
H28117-06
AAH28117-03EA
95.8
USD
AAH28117-03
AAH28117-MD
AAH28117-06
1-Benzyloxycarbonylpyrrolidine-3-carboxaldehyde 97%
1-Benzyloxycarbonylpyrrolidine-3-carboxaldehyde
Purity: 97%
CAS No.: 276872-86-7
Molecular Formula: C13H15NO3
Formula Weight: 233.27
Storage Temperature: 2°C to 8°C
MDL No.: MFCD05861538
Formula:
C₁₃H₁₅NO₃ MW: 233.27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05861538 CAS Number: 276872-86-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...