1,2-Bis(4-methoxyphenyl)-1,1,2,2-tetramethyldisilane 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Bis(4-methoxyphenyl)-1,1,2,2-tetramethyldisilane

H28047-03 H28047-06
AAH28047-03EA 69.2 USD
AAH28047-03 AAH28047-06
1,2-Bis(4-methoxyphenyl)-1,1,2,2-tetramethyldisilane 97%
1,2-Bis(4-methoxyphenyl)-1,1,2,2-tetramethyldisilane

Purity: 97% CAS No.: 6009-50-3 Molecular Formula: C18H26O2Si2 Formula Weight: 330.57 MDL No.: MFCD09266190

Formula: C₁₈H₂₆O₂Si₂
MW: 330.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09266190
CAS Number: 6009-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR