2-Cyclohexylethylamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(2-Aminoethyl)cyclohexane, 2-Cyclohexylethylamine
H27987-03
H27987-06
AAH27987-03EA
120
USD
AAH27987-03
AAH27987-06
2-Cyclohexylethylamine 97%
2-Cyclohexylethanamine
Purity: 97%
CAS No.: 4442-85-7
Molecular Formula: C8H17N
Formula Weight: 127.23
UN No.: 2735
MDL No.: MFCD00058668
Formula:
C₈H₁₇N Storage Temperature: Ambient |
MDL Number:
MFCD01734731 CAS Number: 4442-85-7 EINECS: 224-673-7 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...