6-Chloro-3-pyridineacetic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(6-Chloropyridin-3-yl)acetic acid

H27966-09 H27966-18
AAH27966-09EA 74.6 USD
AAH27966-09 AAH27966-18
6-Chloro-3-pyridineacetic acid 97%
6-Chloro-3-pyridineacetic acid

Purity: 97% CAS No.: 39891-13-9 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Melting Point: 172-176° MDL No.: MFCD01863172

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Melting Pt: 172…176 °C
Storage Temperature: Ambient
MDL Number: MFCD01863172
CAS Number: 39891-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR