5-Aminopyridin-2(1H)-one 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Amino-2-hydroxypyridine, 3-Amino-6-hydroxypyridine, 5-Amino-2(1H)-pyridone
H27961-06
H27961-03
AAH27961-06EA
263
USD
AAH27961-06
AAH27961-03
5-Aminopyridin-2(1H)-one 97%
5-Aminopyridin-2(1H)-one
Purity: 97%
CAS No.: 33630-94-3
Molecular Formula: C5H6N2O
Formula Weight: 110.12
Storage Temperature: 2°C to 8°C
Melting Point: ca 180° dec.
MDL No.: MFCD03427652
Formula:
C₅H₆N₂O MW: 110.12 g/mol Melting Pt: ∼ 180 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD03427652 CAS Number: 33630-94-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...