4-(2-Methoxyethylaminocarbonyl)benzeneboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-(2-Methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
H27945-03
H27945-06
AAH27945-03EA
61.5
USD
AAH27945-03
AAH27945-06
4-(2-Methoxyethylaminocarbonyl)benzeneboronic acid pinacol ester 97%
4-(2-Methoxyethylaminocarbonyl)benzeneboronic acid pinacol ester
Purity: 97%
CAS No.: 1073353-60-2
Molecular Formula: C16H24BNO4
Formula Weight: 305.18
Melting Point: 73-76°
MDL No.: MFCD09266188
Formula:
C₁₆H₂₄BNO₄ MW: 305.18 g/mol Melting Pt: 73…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD09266188 CAS Number: 1073353-60-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...