(3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: (3R,4S,5R,6R)-3,4,5-tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol, 2,3,4,6-Tetra-O-benzyl-D-glucopyranose

H27888-03 H27888-06
AAH27888-03EA 50 USD
AAH27888-03 AAH27888-06
(3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol ≥98%
(3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol

Purity: 98+% CAS No.: 4132-28-9 Molecular Formula: C34H36O6 Formula Weight: 540.65 Storage Temperature: -30°C to -10°C Melting Point: 150-155° MDL No.: MFCD00066004

Formula: C₃₄H₃₆O₆
MW: 540.65 g/mol
Melting Pt: 150…155 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066004
CAS Number: 4132-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR