N-Boc-D-alaninol 98%, ee 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Boc-D-Alaninol, (R)-2-(BOC-amino)-1-propanol
H27887-06
H27887-03
H27887-14
AAH27887-06EA
113
USD
AAH27887-06
AAH27887-03
AAH27887-14
N-Boc-D-alaninol 98%, ee 98%
N-Boc-D-alaninol
Purity: 98%
CAS No.: 106391-86-0
Molecular Formula: C8H17NO3
Formula Weight: 175.23
Melting Point: 58-62°
Optical Rotation: +11° (c=1 in chloroform)
Formula:
C₈H₁₇NO₃ MW: 175.2 g/mol Melting Pt: 58…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00235912 CAS Number: 106391-86-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...