1-tert-Butyl-3-methylpiperazine-1,3-dicarboxylate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate

H27886-03 H27886-06
AAH27886-03EA 157 USD
AAH27886-03 AAH27886-06
1-tert-Butyl-3-methylpiperazine-1,3-dicarboxylate
1-tert-Butyl-3-methylpiperazine-1,3-dicarboxylate

CAS No.: 129799-08-2 Molecular Formula: C11H20N2O4 Formula Weight: 244.29 MDL No.: MFCD01632465

Formula: C₁₁H₂₀N₂O₄
MW: 244.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01632465
CAS Number: 129799-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR