Trimethoxy(pentamethylcyclopentadienyl)titanium(IV) ≥97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pentamethylcyclopentadienyltitanium trimethoxide

H27885-03 H27885-06
AAH27885-03EA 120 USD
AAH27885-03 AAH27885-06
Trimethoxy(pentamethylcyclopentadienyl)titanium(IV) ≥97%
Trimethoxy(pentamethylcyclopentadienyl)titanium(IV)

Purity: 97+% CAS No.: 123927-75-3 Molecular Formula: C13H24O3Ti Formula Weight: 276.21 Flash Point: 61°(142°F) Refractive Index: 1.554 Boiling Point: 285° MDL No.: MFCD00269850

Formula: C₁₃H₂₄O₃Ti
MW: 276.199 g/mol
Boiling Pt: 285 °C
Density: 1.081
Flash Pt: 61 °C (142 °F)
Storage Temperature: Ambient
MDL Number: MFCD00269850
CAS Number: 123927-75-3
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR