tert-Butoxybis(dimethylamino)methane 90%, tech.

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bredereck's reagent, 1-tert-Butoxy-N,N,N',N'-tetramethylmethylenenediamine, [(tert-butoxy)(dimethylamino)methyl]dimethylamine, tert-Butoxy bis(dimethylamino)methane

H27861-03 H27861-06 H27861-14
AAH27861-03EA 48.2 USD
AAH27861-03 AAH27861-06 AAH27861-14
tert-Butoxybis(dimethylamino)methane 90%, tech.
tert-Butoxybis(dimethylamino)methane

Purity: 90% CAS No.: 5815-08-7 Molecular Formula: C9H22N2O Formula Weight: 174.30 Flash Point: 41°(106°F) Refractive Index: 1.422 Boiling Point: 50-55°/15mm UN No.: 1993 MDL No.: MFCD00042858

Formula: C₉H₂₂N₂O
MW: 174.288 g/mol
Boiling Pt: 50…55 °C/15 mm
Density: 0.844
Flash Pt: 41 °C (106 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042858
CAS Number: 5815-08-7
EINECS: 227-383-9
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR