Dichloro(dimethylamino)phosphine 98%

Supplier: Thermo Scientific Chemicals
Danger

H27856-06 H27856-14
AAH27856-06EA 68.68 USD
AAH27856-06 AAH27856-14
Dichloro(dimethylamino)phosphine 98%
Dichloro(dimethylamino)phosphine

Purity: 98% CAS No.: 683-85-2 Molecular Formula: C2H6Cl2NP Formula Weight: 145.96 Storage Temperature: 2°C to 8°C Flash Point: -23°(-9°F) Boiling Point: 150° UN No.: 2924 MDL No.: MFCD00013613

Formula: C₂H₆Cl₂NP
Boiling Pt: 150 °C
Density: 1.271
Flash Pt: –23 °C (–9 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013613
CAS Number: 683-85-2
UN: 2924
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR