(E)-5-(2-Bromovinyl)-2'-deoxyuridine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 5-((E)-2-Bromovinyl)-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, Brivudine, BVdU

H27853-MF H27853-MD H27853-03
AAH27853-MFEA 82.61 USD
AAH27853-MF AAH27853-MD AAH27853-03
(E)-5-(2-Bromovinyl)-2'-deoxyuridine 98%
(E)-5-(2-Bromovinyl)-2'-deoxyuridine

Purity: 98% CAS No.: 69304-47-8 Molecular Formula: C11H13BrN2O5 Formula Weight: 333.14 Storage Temperature: 2°C to 8°C Melting Point: ca 165° dec. Optical Rotation: +10° (c=1 in methanol) MDL No.: MFCD00058585

Formula: C₁₁H₁₃BrN₂O₅
MW: 333.14 g/mol
Melting Pt: ∼ 165 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00058585
CAS Number: 69304-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR