7,8-Dihydroxyflavone hydrate 97%
Supplier: Thermo Scientific Chemicals
H27852-03
H27852-06
AAH27852-03EA
122
USD
AAH27852-03
AAH27852-06
7,8-Dihydroxyflavone hydrate 97%
7,8-Dihydroxyflavone hydrate
Purity: 97%
CAS No.: 38183-03-8
Molecular Formula: C15H10O4.xH2O
Formula Weight: 254.24(anhy)
Melting Point: 250-252°
MDL No.: MFCD00006836
Formula:
C₁₅H₁₀O₄ Melting Pt: 250…252 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006836 CAS Number: 38183-03-8 EINECS: 253-812-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...