N-Boc-cis-4-hydroxy-L-proline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-BOC-cis-4-Hydroxy-L-proline, cis-N-(tert-Butoxycarbonyl)-4-hydroxy-L-proline, Boc-cis-Hyp-OH, cis-N-Boc-4-hydroxy-L-proline, Boc-Cis-Hyp-OH, (2S,4S)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid
H27850-03
H27850-06
AAH27850-03EA
58.3
USD
AAH27850-03
AAH27850-06
AAH27850-MD
N-Boc-cis-4-hydroxy-L-proline 97%
N-Boc-cis-4-hydroxy-L-proline
Purity: 97%
CAS No.: 87691-27-8
Molecular Formula: C10H17NO5
Formula Weight: 231.25
Melting Point: ca 146° dec.
MDL No.: MFCD02094406
Formula:
C₁₀H₁₇NO₅ MW: 231.248 g/mol Melting Pt: ca 146 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD02094406 CAS Number: 87691-27-8 EINECS: 627-315-1 UN: 3077 ADR: "9",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...