1-Hexylboronic acid pinacol ester 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H27831-03 H27831-06 H27831-14
AAH27831-03EA 41.5 USD
AAH27831-03 AAH27831-06 AAH27831-14
1-Hexylboronic acid pinacol ester 98%
1-Hexylboronic acid pinacol ester

Purity: 98% CAS No.: 86308-26-1 Molecular Formula: C12H25BO2 Formula Weight: 212.14 Flash Point: 84°(183°F) Refractive Index: 1.426 MDL No.: MFCD08457665

Formula: C₁₂H₂₅BO₂
Density: 1.167
Flash Pt: 84 °C(183 °F)
Storage Temperature: Ambient
MDL Number: MFCD08457665
CAS Number: 86308-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR