4-Phenyl-3-butyn-2-one 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27826-03
H27826-06
AAH27826-03EA
60.2
USD
AAH27826-03
AAH27826-06
4-Phenyl-3-butyn-2-one 96%
4-Phenyl-3-butyn-2-one
Purity: 96%
CAS No.: 1817-57-8
Molecular Formula: C10H8O
Formula Weight: 144.17
Storage Temperature: 2°C to 8°C
Flash Point: 95°(203°F)
Refractive Index: 1.5740
Boiling Point: 75-76°/0.8mm
MDL No.: MFCD00008776
Formula:
C₁₀H₈O MW: 144.17 g/mol Boiling Pt: 75…76 °C/0.8 mm Density: 0.990 Flash Pt: 95 °C(203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008776 CAS Number: 1817-57-8 EINECS: 217-327-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...