1-Boc-3-oxopiperazine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-butyl 3-oxopiperazine-1-carboxylate, tert-butyl 3-oxo-1-piperazinecarboxylate, 1-Boc-3-Oxopiperazine, 4-N-BOC-2-oxo-piperazine, 4-Boc-2-Oxopiperazine

H27823-03 H27823-06
AAH27823-03EA 70.7 USD
AAH27823-03 AAH27823-06
1-Boc-3-oxopiperazine 98%
1-Boc-3-oxopiperazine

Purity: 98% CAS No.: 76003-29-7 Molecular Formula: C9H16N2O3 Formula Weight: 200.23 Melting Point: 156-160° MDL No.: MFCD02181069

Formula: C₉H₁₆N₂O₃
MW: 200.238 g/mol
Melting Pt: 156…160 °C
Storage Temperature: Ambient
MDL Number: MFCD02181069
CAS Number: 76003-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR