N-Cbz-D-phenylalaninol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-Benzyloxycarbonyl-D-phenylalaninol, Z-D-Phenylalaninol, Cbz-D-Phenylalaninol
H27819-03
H27819-06
AAH27819-03EA
26.2
USD
AAH27819-03
AAH27819-14
AAH27819-06
N-Cbz-D-phenylalaninol 98%
N-Cbz-D-phenylalaninol
Purity: 98%
CAS No.: 58917-85-4
Molecular Formula: C17H19NO3
Formula Weight: 285.35
Melting Point: 92-95°
Optical Rotation: 42° (c=1 in methanol)
Formula:
C₁₇H₁₉NO₃ Melting Pt: 92…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191193 CAS Number: 58917-85-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...