3-Bromo-p-toluonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

H27806-06 H27806-14
AAH27806-06EA 31.2 USD
AAH27806-06 AAH27806-14
3-Bromo-p-toluonitrile 97%
3-Bromo-p-toluonitrile

Purity: 97% CAS No.: 42872-74-2 Molecular Formula: C8H6BrN Formula Weight: 196.05 Flash Point: 110°(230°F) Melting Point: 39-43° UN No.: 3439 MDL No.: MFCD06797818

Formula: C₈H₆BrN
Melting Pt: 39…43 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD06797818
CAS Number: 42872-74-2
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR