6-Nitro-1,3-benzoxazol-2(3H)-one 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Nitrobenzoxazole-2(3H)-one
H27796-06
H27796-03
AAH27796-06EA
79.2
USD
AAH27796-06
AAH27796-03
6-Nitro-1,3-benzoxazol-2(3H)-one 98%
6-Nitro-1,3-benzoxazol-2(3H)-one
Purity: 98%
CAS No.: 4694-91-1
Molecular Formula: C7H4N2O4
Formula Weight: 180.12
Melting Point: 244-249°
MDL No.: MFCD00463755
Formula:
C₇H₄N₂O₄ MW: 180.12 g/mol Melting Pt: 244…249 °C Storage Temperature: Ambient |
MDL Number:
MFCD00463755 CAS Number: 4694-91-1 EINECS: 225-157-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...