1,3-Dimethylimidazolium dimethylphosphate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3-Dimethylimidazolium Dimethyl Phosphate

H27788-06 H27788-18
AAH27788-06EA 42.8 USD
AAH27788-06 AAH27788-18
1,3-Dimethylimidazolium dimethylphosphate 98%
1,3-Dimethylimidazolium dimethylphosphate

Purity: 98% CAS No.: 654058-04-5 Molecular Formula: C7H15N2O4P Formula Weight: 222.18 MDL No.: MFCD08275369

Formula: C₇H₁₅N₂O₄P
MW: 222.18 g/mol
Density: 1.277
Storage Temperature: Ambient
MDL Number: MFCD08275369
CAS Number: 654058-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR