2-Nitroterephthalic acid-4-methyl ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27779-06
H27779-14
AAH27779-06EA
87.2
USD
AAH27779-06
AAH27779-14
2-Nitroterephthalic acid-4-methyl ester 97%
2-Nitroterephthalic acid 4-methyl ester
Purity: 97%
CAS No.: 55737-66-1
Molecular Formula: C9H7NO6
Formula Weight: 225.17
Melting Point: 132-136°
MDL No.: MFCD06203344
Formula:
C₉H₇NO₆ Melting Pt: 132…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD06203344 CAS Number: 55737-66-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...