1-n-Butyl-4-[(4-butylphenyl)ethynyl]benzene ≥99%

Supplier: Thermo Scientific Chemicals

H27776-03 H27776-09
AAH27776-03EA 55.2 USD
AAH27776-03 AAH27776-09
1-n-Butyl-4-[(4-butylphenyl)ethynyl]benzene ≥99%
1-n-Butyl-4-[(4-butylphenyl)ethynyl]benzene

Purity: 99+% CAS No.: 80221-11-0 Molecular Formula: C22H26 Formula Weight: 290.45 Melting Point: 40-42° Boiling Point: 172-174°/0.5mm MDL No.: MFCD04038794

Formula: C₂₂H₂₆
MW: 290.45 g/mol
Boiling Pt: 172…174 °C/0.5 mm
Melting Pt: 40…42 °C
Storage Temperature: Ambient
MDL Number: MFCD04038794
CAS Number: 80221-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR