Ethyl-2-hydroxy-6-methylpyridine-4-carboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 2-hydroxy-6-methylisonicotinate
H27775-03
H27775-06
AAH27775-MDEA
52.49
USD
AAH27775-MD
AAH27775-03
AAH27775-06
Ethyl-2-hydroxy-6-methylpyridine-4-carboxylate 97%
Ethyl-2-hydroxy-6-methylpyridine-4-carboxylate
Purity: 97%
CAS No.: 150190-03-7
Molecular Formula: C9H11NO3
Formula Weight: 181.20
Melting Point: 188-190°
MDL No.: MFCD09953494
Formula:
C₉H₁₁NO₃ MW: 181.20 g/mol Melting Pt: 188…190 °C Storage Temperature: Ambient |
MDL Number:
MFCD09953494 CAS Number: 150190-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...