Methyl hydrogen malonate 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27772-06
H27772-14
H27772-22
AAH27772-06EA
62.5
USD
AAH27772-06
AAH27772-14
AAH27772-22
Methyl hydrogen malonate 96%
Methyl hydrogen malonate
Purity: 96%
CAS No.: 16695-14-0
Molecular Formula: HO2CCH2CO2CH3
Formula Weight: 118.09
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4295
MDL No.: MFCD00667796
Formula:
HO₂CCH₂CO₂CH₃ Density: 1.128 Storage Temperature: Ambient |
MDL Number:
MFCD00667796 CAS Number: 16695-14-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...