7-(Trifluoromethyl)quinoline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27771-03
H27771-MD
AAH27771-03EA
166
USD
AAH27771-03
AAH27771-MD
7-(Trifluoromethyl)quinoline 97%
7-(Trifluoromethyl)quinoline
Purity: 97%
CAS No.: 325-14-4
Molecular Formula: C10H6F3N
Formula Weight: 197.16
Melting Point: 65-66°
MDL No.: MFCD00833760
Formula:
C₁₀H₆F₃N Melting Pt: 65…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00833760 CAS Number: 325-14-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...