Methyl-4-methoxyindole-2-carboxylate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 4-methoxy-1H-indole-2-carboxylate
H27743-MD
H27743-03
AAH27743-MDEA
45
USD
AAH27743-MD
AAH27743-03
Methyl-4-methoxyindole-2-carboxylate 99%
Methyl-4-methoxyindole-2-carboxylate
Purity: 99%
CAS No.: 111258-23-2
Molecular Formula: C11H11NO3
Formula Weight: 205.21
Melting Point: 140-142°
MDL No.: MFCD00134300
Formula:
C₁₁H₁₁NO₃ MW: 205.21 g/mol Melting Pt: 140…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134300 CAS Number: 111258-23-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...