1-Cyanocyclopropanecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

H27730-03 H27730-06
AAH27730-03EA 37 USD
AAH27730-03 AAH27730-06
1-Cyanocyclopropanecarboxylic acid 97%
1-Cyanocyclopropanecarboxylic acid

Purity: 97% CAS No.: 6914-79-0 Molecular Formula: C5H5NO2 Formula Weight: 111.10 Melting Point: 143-149° UN No.: 2923 MDL No.: MFCD00190651

Formula: C₅H₅NO₂
Melting Pt: 143…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00190651
CAS Number: 6914-79-0
UN: 2923
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR