(R)-(+)-2-Methyl-2-propanesulfinamide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(R)-2-methylpropane-2-sulphinamide, R)-(+)-tert-Butyl sulphinamide, (R)-2-Methyl-2-propanesulphinamide, (R)-(+)-tert-Butylsulphinamide, (R)-(+)-t-Butylsulphinamide
H27724-06
H27724-03
H27724-14
AAH27724-06EA
227
USD
AAH27724-06
AAH27724-03
AAH27724-14
(R)-(+)-2-Methyl-2-propanesulfinamide 98%
(R)-(+)-2-Methyl-2-propanesulfinamide
Purity: 98%
CAS No.: 196929-78-9
Molecular Formula: C4H11NOS
Formula Weight: 121.20
Storage Temperature: 2°C to 8°C
Melting Point: ca 100-105°
MDL No.: MFCD05861479
Formula:
C₄H₁₁NOS MW: 121.20 g/mol Melting Pt: ∼ 100…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD05861479 CAS Number: 196929-78-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...