4-Cyanopiperidine 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Piperidine-4-carbonitrile
H27722-06
H27722-14
AAH27722-06EA
71.3
USD
AAH27722-06
AAH27722-14
4-Cyanopiperidine 96%
4-Cyanopiperidine
Purity: 96%
CAS No.: 4395-98-6
Molecular Formula: C6H10N2
Formula Weight: 110.16
Flash Point: 91°(196°F)
Refractive Index: 1.4755
Odor: Amine-like
Boiling Point: 226°
UN No.: 3276
MDL No.: MFCD05022468
Formula:
C₆H₁₀N₂ MW: 110.16 g/mol Boiling Pt: 226 °C Density: 0.98 Flash Pt: 91 °C (196 °F) Storage Temperature: Ambient |
MDL Number:
MFCD05022468 CAS Number: 4395-98-6 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...