5-Bromo-1,4-benzodioxane 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromo-2,3-dihydro-1,4-benzodioxane

H27709-03 H27709-MD
AAH27709-03EA 218 USD
AAH27709-03 AAH27709-MD
5-Bromo-1,4-benzodioxane 95%
5-Bromo-1,4-benzodioxane

Purity: 95% CAS No.: 58328-39-5 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 Refractive Index: 1.5955

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095029
CAS Number: 58328-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR