3-Iodo-1-(phenylsulfonyl)indole 95%
Supplier: Thermo Scientific Chemicals
H27696-03
H27696-MD
AAH27696-03EA
139
USD
AAH27696-03
AAH27696-MD
3-Iodo-1-(phenylsulfonyl)indole 95%
3-Iodo-1-(phenylsulfonyl)indole
Purity: 95%
CAS No.: 80360-14-1
Molecular Formula: C14H10INO2S
Formula Weight: 383.20
Melting Point: 125-129°
MDL No.: MFCD09037474
Formula:
C₁₄H₁₀INO₂S Melting Pt: 125…129 °C Storage Temperature: Ambient |
MDL Number:
MFCD09037474 CAS Number: 80360-14-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...