(R)-(+)-1,2,3,4-Tetrahydro-1-naphthoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-1,2,3,4-Tetrahydronaphthoic acid

H27673-06 H27673-MD H27673-03
AAH27673-06EA 574 USD
AAH27673-06 AAH27673-MD AAH27673-03
(R)-(+)-1,2,3,4-Tetrahydro-1-naphthoic acid 98%
(R)-(+)-1,2,3,4-Tetrahydro-1-naphthoic acid

Purity: 98% CAS No.: 23357-47-3 Molecular Formula: C11H12O2 Formula Weight: 176.22 Melting Point: 56-58° Optical Rotation: +12° (c=1 in ethanol)

Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Melting Pt: 56…58 °C
Storage Temperature: Ambient
MDL Number: MFCD07368364
CAS Number: 23357-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR