Dimethylamine 2 M in THF
Supplier: Thermo Scientific Chemicals
|
Danger
|
H27665-AE
H27665-AU
AAH27665-AEEA
125
USD
AAH27665-AE
AAH27665-AU
Dimethylamine 2 M in THF
Dimethylamine
CAS No.: 124-40-3
Molecular Formula: C2H7N
Formula Weight: 45.08
Flash Point: -36°(-33°F)
Boiling Point: ca 60°
Beilstein Reference: 605257
Merck Reference: 14,3228
UN No.: 2924
MDL No.: MFCD00008288
Formula:
C₂H₇N Boiling Pt: ∼ 60 °C Density: 0.85 Flash Pt: –36 °C (–33 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008288 CAS Number: 124-40-3 EINECS: 204-697-4 UN: 2924 ADR: 3,II Merck Index: 14,3228 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...