Cyclopropylboronic acid pinacol ester 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
H27642-03
H27642-06
AAH27642-03EA
86
USD
AAH27642-03
AAH27642-06
Cyclopropylboronic acid pinacol ester 96%
Cyclopropylboronic acid pinacol ester
Purity: 96%
CAS No.: 126689-01-8
Molecular Formula: C9H17BO2
Formula Weight: 168.04
Storage Temperature: -30°C to -10°C
Flash Point: 40°(104°F)
Boiling Point: 146°
UN No.: 3272
MDL No.: MFCD05663847
Formula:
C₉H₁₇BO₂ Boiling Pt: 146 °C Density: 0.922 Flash Pt: 40 °C(104 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD05663847 CAS Number: 126689-01-8 UN: 3272 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...