5'-m-Terphenylboronic Acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: [1,1':3',1''-Terphenyl]-5'-ylboronic acid

H27640-MD H27640-03 H27640-06
AAH27640-MDEA 26.9 USD
AAH27640-MD AAH27640-03 AAH27640-06
5'-m-Terphenylboronic Acid 95%
5'-m-Terphenylboronic acid

Purity: 95% CAS No.: 128388-54-5 Molecular Formula: C18H15BO2 Formula Weight: 274.13 Melting Point: 292-296° MDL No.: MFCD09953491

Formula: C₁₈H₁₅BO₂
Melting Pt: 292…296 °C
Storage Temperature: Ambient
MDL Number: MFCD09953491
CAS Number: 128388-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR