D-Homophenylalanine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (R)-2-Amino-4-phenylbutyric Acid, H-D-HomoPhe-OH, H-D-HoPhe-OH

H27629-03 H27629-06
AAH27629-03EA 76 USD
AAH27629-03 AAH27629-06
D-Homophenylalanine 98%
D-Homophenylalanine

Purity: 98% CAS No.: 82795-51-5 Molecular Formula: C10H13NO2 Formula Weight: 179.22 Melting Point: 300° Beilstein Reference: 4675530 MDL No.: MFCD00063091

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00063091
CAS Number: 82795-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR