4-(Methoxycarbonyl)phenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-(Methoxycarbonyl)phenyl)boronic acid, 4-(Methoxycarbonyl)benzeneboronic acid

H27627-03 H27627-06
AAH27627-03EA 68.2 USD
AAH27627-03 AAH27627-06
4-(Methoxycarbonyl)phenylboronic acid 97%
4-(Methoxycarbonyl)phenylboronic acid

Purity: 97% CAS No.: 99768-12-4 Molecular Formula: C8H9BO4 Formula Weight: 179.97 Melting Point: 220-225° MDL No.: MFCD01632203

Formula: C₈H₉BO₄
MW: 179.97 g/mol
Melting Pt: 220…225 °C
Storage Temperature: Ambient
MDL Number: MFCD01632203
CAS Number: 99768-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR