1-(1-Methylpiperidin-4-yl)piperazine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-(1-Methyl-4-piperidinyl)piperazine
H27624-03
H27624-06
AAH27624-03EA
47.2
USD
AAH27624-03
AAH27624-06
1-(1-Methylpiperidin-4-yl)piperazine 98%
1-(1-Methylpiperidin-4-yl)piperazine
Purity: 98%
CAS No.: 23995-88-2
Molecular Formula: C10H21N3
Formula Weight: 183.30
Refractive Index: 1.508
Melting Point: 29-32°
UN No.: 3259
MDL No.: MFCD01075186
Formula:
C₁₀H₂₁N₃ MW: 183.3 g/mol Melting Pt: 29…32 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075186 CAS Number: 23995-88-2 UN: 2735 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...