2H-1,4-Benzoxazin-3(4H)-one 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27622-06
H27622-14
AAH27622-06EA
31.1
USD
AAH27622-06
AAH27622-14
2H-1,4-Benzoxazin-3(4H)-one 99%
2H-1,4-Benzoxazin-3(4H)-one
Purity: 99%
CAS No.: 5466-88-6
Molecular Formula: C8H7NO2
Formula Weight: 149.15
Melting Point: 173-175°
MDL No.: MFCD00158536
Formula:
C₈H₇NO₂ Melting Pt: 173…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00158536 CAS Number: 5466-88-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...