Methyl-3-amino-5,6-dichloropyrazine-2-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-amino-5,6-dichloropyrazine-2-carboxylate

H27614-03 H27614-06
AAH27614-03EA 77 USD
AAH27614-03 AAH27614-06
Methyl-3-amino-5,6-dichloropyrazine-2-carboxylate 97%
Methyl 3-amino-5,6-dichloropyrazine-2-carboxylate

Purity: 97% CAS No.: 1458-18-0 Molecular Formula: C6H5Cl2N3O2 Formula Weight: 222.03 Melting Point: 230-233° MDL No.: MFCD00010431

Formula: C₆H₅Cl₂N₃O₂
MW: 222.03 g/mol
Melting Pt: 230…233 °C
Storage Temperature: Ambient
MDL Number: MFCD00010431
CAS Number: 1458-18-0
EINECS: 215-947-7
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR