(S)-(-)-1,2,3,4-Tetrahydro-1-naphthoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-1,2,3,4-Tetrahydro-1-naphthoic acid, (1S)-1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid
H27612-03
H27612-MD
AAH27612-03EA
88.6
USD
AAH27612-03
AAH27612-06
AAH27612-MD
(S)-(-)-1,2,3,4-Tetrahydro-1-naphthoic acid 98%
(S)-(-)-1,2,3,4-Tetrahydro-1-naphthoic acid
Purity: 98%
CAS No.: 85977-52-2
Molecular Formula: C11H12O2
Formula Weight: 176.22
Melting Point: 55-57°
Optical Rotation: -12° (c=1 in ethanol)
Formula:
C₁₁H₁₂O₂ MW: 176.22 g/mol Melting Pt: 55…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD07368365 CAS Number: 85977-52-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...