1-Benzyl-N-methyl-4-piperidinamine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Methylamino)-1-benzylpiperidine

H27606-03 H27606-06
AAH27606-03EA 34.7 USD
AAH27606-03 AAH27606-06 AAH27606-14
1-Benzyl-N-methyl-4-piperidinamine 98%
1-Benzyl-N-methyl-4-piperidinamine

Purity: 98% CAS No.: 7006-50-0 Molecular Formula: C13H20N2 Formula Weight: 204.31 MDL No.: MFCD03931054

Formula: C₁₃H₂₀N₂
MW: 204.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03931054
CAS Number: 7006-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR