7-Bromo-3,4-dihydro-2H-1,5-benzodioxepine 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27594-03
H27594-06
AAH27594-03EA
45.1
USD
AAH27594-03
AAH27594-06
AAH27594-14
7-Bromo-3,4-dihydro-2H-1,5-benzodioxepine 96%
7-Bromo-3,4-dihydro-2H-1,5-benzodioxepine
Purity: 96%
CAS No.: 147644-11-9
Molecular Formula: C9H9BrO2
Formula Weight: 229.07
Flash Point:
Formula:
C₉H₉BrO₂ Boiling Pt: 145…148 °C/16 mm Flash Pt: >120 °C(248 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00508291 CAS Number: 147644-11-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...