5-Aminopicolinic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Aminopyridine-2-carboxylic acid, 5-Amino-2-pyridinecarboxylic acid
H27589-03
H27589-06
AAH27589-03EA
112
USD
AAH27589-03
AAH27589-06
5-Aminopicolinic acid 98%
5-Aminopicolinic acid
Purity: 98%
CAS No.: 24242-20-4
Molecular Formula: C6H6N2O2
Formula Weight: 138.13
Odor: Odorless
Melting Point: 217-219°
MDL No.: MFCD02684600
Formula:
C₆H₆N₂O₂ MW: 138.13 g/mol Melting Pt: 217…219 °C Storage Temperature: Ambient |
MDL Number:
MFCD02684600 CAS Number: 24242-20-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...