3-(1H-Tetrazol-5-yl)benzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(1H-Tetrazol-5-yl)benzoic Acid

H27581-03 H27581-MD
AAH27581-03EA 91.3 USD
AAH27581-03 AAH27581-MD
3-(1H-Tetrazol-5-yl)benzoic acid 97%
3-(1H-Tetrazol-5-yl)benzoic acid

Purity: 97% CAS No.: 73096-39-6 Molecular Formula: C8H6N4O2 Formula Weight: 190.16 Melting Point: 284-286° MDL No.: MFCD03821131

Formula: C₈H₆N₄O₂
MW: 190.16 g/mol
Melting Pt: 284…286 °C
Storage Temperature: Ambient
MDL Number: MFCD03821131
CAS Number: 73096-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR