4-N-Boc-amino-4-carboxytetrahydropyran 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-((tert-Butoxycarbonyl)amino)tetrahydro-2H-pyran-4-carboxylic acid

H27578-03 H27578-MD
AAH27578-03EA 207 USD
AAH27578-03 AAH27578-MF AAH27578-MD
4-N-Boc-amino-4-carboxytetrahydropyran 95%
4-N-Boc-amino-4-carboxytetrahydropyran

Purity: 95% CAS No.: 172843-97-9 Molecular Formula: C11H19NO5 Formula Weight: 245.27 Melting Point: 157° dec. MDL No.: MFCD02683136

Formula: C₁₁H₁₉NO₅
MW: 245.275 g/mol
Melting Pt: 157 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD02683136
CAS Number: 172843-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR