(R,R)-(-)-2,5-Bis(methoxymethyl)pyrrolidine 97%

Supplier: Thermo Scientific Chemicals
Warning

H27577-ME H27577-MC
AAH27577-MEEA 331 USD
AAH27577-ME AAH27577-MC
(R,R)-(-)-2,5-Bis(methoxymethyl)pyrrolidine 97%
(R,R)-(-)-2,5-Bis(methoxymethyl)pyrrolidine

Purity: 97% CAS No.: 90290-05-4 Molecular Formula: C8H17NO2 Formula Weight: 159.23 Refractive Index: 1.4490 Boiling Point: 106-108°/18mm Optical Rotation: -8° (c=3 in ethanol) MDL No.: MFCD00210015

Formula: C₈H₁₇NO₂
MW: 159.23 g/mol
Boiling Pt: 106…108 °C/18 mm
Storage Temperature: Ambient
MDL Number: MFCD00210015
CAS Number: 90290-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR